Vancomycin Hydrochlorid (aus Streptomyces lysosup)

Lieferant: Enzo Life Sciences

ALX380279G001 ALX380279M250
ENZOALX380279G001EA 286 EUR
ENZOALX380279G001 ENZOALX380279M250
Vancomycin Hydrochlorid (aus Streptomyces lysosup)
Vancomycin Hydrochlorid

Glycopeptide antibiotic. Interferes with cell wall synthesis. Effective against Gram-positive bacteria.


White to light brown powder


Soluble in water (50 mg/ml)


Verpackung: Shipping temperature: Ambient

Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD03613611
CAS-Nummer: 1404-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR