Ciprofloxacin ≥98%
Lieferant: ENZO LIFE SCIENCES
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.
- Effective against Mycobacterium tuberculosis
- Potentially anxiogenic
White to off-white powder
Soluble in dilute hydrochloric acid; insoluble in water
Verpackung: Shipping temperature: Ambient
Formel:
C₁₇H₁₈FN₃O₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00185755 CAS-Nummer: 85721-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...