Rufloxacin ≥98%
Lieferant: ENZO LIFE SCIENCES
ALX380300M250
ENZOALX380300M250EA
373
EUR
ENZOALX380300M250
Rufloxacin ≥98%
Rufloxacin
Fluoroquinolone antibacterial agent. Structurally similar to ofloxacin (Prod. No. ALX-380-297). Specific inhibitor of bacterial DNA gyrase (topoisomerase). Inhibits B-cell differentiation. Shows photosensitizing effects.
Formel:
C₁₇H₁₈FN₃O₃S·HCl Molecular Weight: 363,4 · 36,5 g/mol |
CAS-Nummer:
106017-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...