Sparfloxacin ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Amino-1-cyclopropyl-7-((3S,5R)-3,5-dimethylpiperazin-1-yl)-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

ALX380302G005
ENZOALX380302G005EA 578 EUR
ENZOALX380302G005
Sparfloxacin ≥98%
Sparfloxacin

Fluoroquinolone with broad spectrum antibacterial activity against Gram-positive and Gram-negative bacteria. Inhibits bacterial DNA gyrase (topoisomerase). Effective against Mycobacterium tuberculosis. Shows photosensitizing effects.

Formel: C₁₉H₂₂F₂N₄O₃
Molecular Weight: 392,407 g/mol
MDL: MFCD00869619
CAS-Nummer: 110871-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR