Tosufloxacin tosylate ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
7-(3-Aminopyrrolidin-1-yl)-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid 4-methylbenzenesulphonic acid salt
ALX380303M250
ENZOALX380303M250EA
157
EUR
ENZOALX380303M250
Tosufloxacin tosylate ≥98% (durch HPLC)
Tosufloxacin tosylate
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Effective against Gram-positive and Gram-negative aerobic bacteria, anaerobic bacteria and Chlamydia trachomatis.
- White to yellow crystalline solid
Slightly soluble in water
Verpackung: Shipping temperature: Ambient
Formel:
C₁₉H₁₅F₃N₄O₃·C₇H₈O₃S Molecular Weight: 404.4 · 172.2 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD01711971 CAS-Nummer: 115964-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...