Enniatin A (aus Fusarium sp.) ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX380310MC05
ENZOALX380310MC05EA 283 EUR
ENZOALX380310MC05
Enniatin A (aus Fusarium sp.) ≥97% (durch HPLC)
Enniatin A

White to off-white crystalline solid


  • Ionophore antibiotic
  • Potential anticancer compound


Soluble in 100% ethanol, methanol, DMSO and dimethyl formamide; almost insoluble in water


Verpackung: Shipping temperature: Ambient

Formel: C₃₆H₆₃N₃O₉
Lagertemperatur: Gefrierschrank
CAS-Nummer: 2503-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR