Quercetin dihydrate ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
Antioxidant flavonoid
Antioxidant flavonoid. Inhibitor of mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases. Inhibitor of protein tyrosine kinases and protein kinase C (PKC). Induces apoptosis. Blocks cells at the G0/G1 interface. Activator of human deacetylase SIRT1. Reversible inhibitor of fatty acid synthase (FAS). Inhibits the production of the inflammatory mediators nitric oxide (NO), TNF-α and IL-12 in activated macrophages.Induces gastric relaxation in mouse stomach, mainly by suppressing Ca²⁺ influx through voltage-dependent calcium channels.
Isolated from Sophora japonica L.
Formel:
C₁₅H₁₄O₉ Molecular Weight: 338,268 g/mol |
MDL:
MFCD00149487 CAS-Nummer: 6151-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...