3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Peonidin chloride

ALX385015M005
ENZOALX385015M005EA 524 EUR
ENZOALX385015M005
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97% (durch HPLC)
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride

Anti-inflammatory and chemopreventive. Antioxidant flavonoid.


Soluble in 100% ethanol or water.

Formel: C₁₆H₁₃O₆Cl
Molecular Weight: 336,73 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00017587
CAS-Nummer: 134-01-0
EINECS: 205-125-6
Merck Index: 14,07143

Bestellen

Spezifikation Testergebnisse

Purity ≥97% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR