Morin ≥85%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2',3,4',5,7-Pentahydroxy-flavone, C.I. 75660, 2-(2,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2′,3,4′,5,7-Pentahydroxyflavone, Aluminum Ionophore I

ALX385016G001
ENZOALX385016G001EA 50.6 EUR
ENZOALX385016G001
Morin ≥85%
Morin

Cell cycle inhibitor


Antioxidant flavonoid. Shows anti-proliferative and antitumor properties. Induces apoptosis. Induces cell cycle arrest at the G2/M phase. Anti-inflammatory compound. Suppresses NF-κB activation. Induces lipid peroxidation and DNA strand breaks.

Formel: C₁₅H₁₀O₇
Molecular Weight: 302,24 g/mol
MDL: MFCD00006826
CAS-Nummer: 480-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR