(+)-Catechin monohydrate ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: (+)-3,3’,4’,5,7-Flavanpentol.H₂O

ALX385017G001
ENZOALX385017G001EA 51.7 EUR
ENZOALX385017G001
(+)-Catechin monohydrate ≥98% (durch HPLC)
(+)-Catechin monohydrate

Free radical scavenger.


Antioxidant flavonoid. Free radical scavenger. Has chemopreventive and antitumor properties.


Soluble in 100% ethanol (50 mg/ml).

Formel: C₁₅H₁₄O₆·H₂O
Molecular Weight: 290,3 g/mol (anhy)
Lagertemperatur: Kühlschrank
MDL: MFCD00075649
CAS-Nummer: 225937-10-0
EINECS: 205-825-1
Merck Index: 14,01902

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR