Delphinidin chloride ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX385028M010
ENZOALX385028M010EA
725
EUR
ENZOALX385028M010
Delphinidin chloride ≥97% (durch HPLC)
Delphinidin chloride
Anthocyanidin with antioxidant effect found in pigmented fruits and vegetables. Shown to inhibit angiogenesis and endothelial cell apoptosis by stimulating nitric oxide (NO) production. Inhibits solar radiation (UVB)-mediated oxidative stress, reducing DNA damage. Delphinidin induces necrosis in hepatocellular carcinoma cells with no caspase activation.
- Antioxidant
Red to dark brown powder.
Soluble in water or DMSO.
Formel:
C₁₅H₁₁O₇Cl Lagertemperatur: Gefrierschrank |
MDL:
MFCD00016663 CAS-Nummer: 528-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...