Nitroprussid-Natrium Dihydrat
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
ALX400001G005
ENZOALX400001G005EA
50.6
EUR
ENZOALX400001G005
Nitroprussid-Natrium Dihydrat
Nitroprussid-Natrium Dihydrat
Formel:
Na₂[Fe(CN)₅NO]·2H₂O Dichte: 1,72 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149192 CAS-Nummer: 13755-38-9 EINECS: 238-373-9 UN: 1588 ADR: 6.1,III Merck Index: 13,08722 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...