Ammonium-1-pyrrolidincarbodithioat ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

ALX400002G005
ENZOALX400002G005EA 49.5 EUR
ENZOALX400002G005
Ammonium-1-pyrrolidincarbodithioat ≥98% (durch HPLC)
Ammonium-1-pyrrolidincarbodithioat

Inhibitor of NF-κB activation (induced by either PMA or TNF-α) in various cell types.


  • Inhibitor of NF-κB activation


White to-off white solid.


Soluble in water.

Formel: C₅H₁₂N₂S₂
Molecular Weight: 146.3 · 18.0 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012720
CAS-Nummer: 5108-96-3
EINECS: 225-834-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR