Natriumdiethyldithiocarbamat Trihydrat ≥99% (durch titrimetrische Analyse)

Lieferant: Enzo Life Sciences
Achtung

ALX400003G025
ENZOALX400003G025EA 86.9 EUR
ENZOALX400003G025
Natriumdiethyldithiocarbamat Trihydrat ≥99% (durch titrimetrische Analyse)
Natriumdiethyldithiocarbamat Trihydrat

Nitric oxide (NO) spin-trapping reagent. Thiol and iron chelator. Inhibits induction of macrophage nitric oxide synthase (NOS). Has been shown to be an inhibitor of the nuclear transcription factor κB (NF-κB).


  • NO spin trapping reagent


White to off-white solid.


Soluble in water (20mg/ml).

Formel: (C₂H₅)₂NCSSNa·3H₂O
Dichte: 1,1 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00150617
CAS-Nummer: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR