5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
ALX400034G001
ALX400034G005
ENZOALX400034G001EA
52.8
EUR
ENZOALX400034G001
ENZOALX400034G005
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Formel:
C₁₄H₈N₂O₈S₂ Schmelzpunkt: 240…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007140 CAS-Nummer: 69-78-3 EINECS: 200-714-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...