Diphenylborsäure(2-aminoethyl)ester (2-APB) 98 HPLC_ASSAY_METHOD

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine

ALX400045M100
ENZOALX400045M100EA 51.7 EUR
ENZOALX400045M100
Diphenylborsäure(2-aminoethyl)ester (2-APB) 98 HPLC_ASSAY_METHOD
Diphenylborsäure(2-aminoethyl)ester (2-APB)

Cell permeable modulator of Ins(1,4,5)-P3-induced Ca²⁺ release. A prototype drug for a group of structurally related calcium channel blockers in human platelets. Has been shown, in the absence of other stimuli, to activate TRPV1, V2 and V3, but not TRPV4, V5 and V6 expressed in HEK 293 cells. Potentiates the effect of capsaicin, protons and heat on TRPV1. It was originally introduced as an IP3 receptor blocker and subsequently as an inhibitor of store-operated Ca²⁺ entry (75µM).


  • TRP channel activator


White to off-white crystalline solid.


Soluble in DMSO (20 mg/ml), dimethyl formamide (20 mg/ml) or 100% ethanol (5 mg/ml); slightly soluble in PBS, pH 7.2 (50 μg/ml).

Formel: C₁₄H₁₆BNO
Molecular Weight: 225.1 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00014823
CAS-Nummer: 524-95-8
EINECS: 208-366-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR