(+)-Indolactam V ≥95% (durch 1H-NMR)

Lieferant: ENZO LIFE SCIENCES
Achtung

ALX420010C300
ENZOALX420010C300EA 204 EUR
ENZOALX420010C300
(+)-Indolactam V ≥95% (durch 1H-NMR)
(+)-Indolactam V

Used as negative control compound in studies with (-)-indolactam V.


Soluble in DMSO or 100% ethanol.

Formel: C₁₇H₂₃N₃O₂
Lagertemperatur: Kühlschrank
CAS-Nummer: 90365-56-3

Bestellen

Spezifikation Testergebnisse

Purity ≥95% (¹H-NMR)
Purity Detail May contain traces of (-)-Indolactam V

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR