GW1929 ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(2-Benzoylphenyl)-L-tyrosine PPARγ agonist

ALX420029M005
ENZOALX420029M005EA 652 EUR
ENZOALX420029M005
GW1929 ≥97% (durch HPLC)
GW1929

PPARγ agonist.


Potent (<10 nM) and subtype-selective (>1000-fold) PPARγ agonist which does not contain a thiazolidinedione moiety. The glucose-lowering effect of GW1929 in rats is 100-fold more potent than that of troglitazone.


Soluble in DMSO or methanol.

Formel: C₃₀H₂₉N₃O₄
Molecular Weight: 495.579 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 196808-24-9

Bestellen

Spezifikation Testergebnisse

Purity ≥97% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR