Carboxy-PTIO potassium salt ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX430001M010 ALX430001M050 ALX430001M250
ENZOALX430001M010EA 84.7 EUR
ENZOALX430001M010 ENZOALX430001M050 ENZOALX430001M250
Carboxy-PTIO potassium salt ≥99% (durch HPLC)
Carboxy-PTIO potassium salt

Water soluble and stable nitric oxide radical scavenger that shows in chemical and biological systems antagonistic action against the free nitric oxide radical (NO·). The inhibitory effects were almost 2-fold stronger than those of NMMA and NOARG. Reacts with nitric oxide in a stoichometric manner. Can be used for an improved detection of nitric oxide in activated macrophage cultures in combination with the Griess reagent. Protects against endotoxin shock.


  • Nitric oxide (NO) scavenger


Blue solid.


Soluble in 100% ethanol, methanol, DMSO or dimethyl formamide. Highly soluble in water or aqueous buffers, e. g. PBS, pH 7.2.

Formel: C₁₄H₁₆KN₂O₄
Molecular Weight: 315.4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00216153
CAS-Nummer: 148819-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR