GEA 5024 ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX430005M010
ENZOALX430005M010EA
114
EUR
ENZOALX430005M010
GEA 5024 ≥97% (durch HPLC)
GEA 5024
Stable nitric oxide donor.
Soluble in water.
Formel:
C₈H₇ClN₄O·HCl Lagertemperatur: Kühlschrank |
CAS-Nummer:
144575-27-9 |
Spezifikation Testergebnisse
Purity | ≥97% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...