(-)-p-Bromotetramisole oxalate ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX430025M005
ENZOALX430025M005EA 117 EUR
ENZOALX430025M005
(-)-p-Bromotetramisole oxalate ≥98% (durch HPLC)
(-)-p-Bromotetramisole oxalate

Potent alkaline phosphatase inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable. 


  • Alkaline phosphatase inhibitor


White powder.


Soluble in water, methanol, DMF or DMSO.

Formel: C₁₁H₁₁BrN₂S·C₂H₂O₄
Lagertemperatur: Kühlschrank
MDL: MFCD00070406
CAS-Nummer: 62284-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR