R59 022 ≥99%
Lieferant: ENZO LIFE SCIENCES
ALX430027M005
ENZOALX430027M005EA 146 EUR
ENZOALX430027M005
R59 022 ≥99%
R59 022
Potentiates PKC activity
Potentiates the activity of protein kinase C (PKC) by inhibiting the diacylglycerol metabolism. Inhibits diacylglycerol kinase without affecting phosphodiesterase or phosphatidyl inositol kinase. It inhibits human platelet DAG kinase in intact cells (IC50=3.6μM) and in isolated membranes (IC50=6.7μM).
CAS-Nummer: 93076-89-2