Demecolcine ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Colcemid
ALX430033M001
ALX430033M005
ENZOALX430033M001EA
63.8
EUR
ENZOALX430033M001
ENZOALX430033M005
Demecolcine ≥98% (durch HPLC)
Demecolcine
Depolymerizes microtubules and inhibits their formation. Induces apoptosis.
Semisynthetic from colchicine (Prod. No. ALX-380-033).
Formel:
C₂₁H₂₅NO₅ Molecular Weight: 371.433 g/mol |
MDL:
MFCD00075459 CAS-Nummer: 477-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...