Demecolcine ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Colcemid

ALX430033M001 ALX430033M005
ENZOALX430033M001EA 63.8 EUR
ENZOALX430033M001 ENZOALX430033M005
Demecolcine ≥98% (durch HPLC)
Demecolcine

Depolymerizes microtubules and inhibits their formation. Induces apoptosis.


Semisynthetic from colchicine (Prod. No. ALX-380-033).

Formel: C₂₁H₂₅NO₅
Molecular Weight: 371.433 g/mol
MDL: MFCD00075459
CAS-Nummer: 477-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR