Probenecid
Lieferant: ENZO LIFE SCIENCES
Synonyme:
4-[(dipropylamino)sulphonyl]benzoic acid
ALX430113G005
ENZOALX430113G005EA
51.7
EUR
ENZOALX430113G005
Probenecid
Probenecid
Formel:
C₁₃H₁₉NO₄S Molecular Weight: 285,4 g/mol |
MDL:
MFCD00038402 CAS-Nummer: 57-66-9 EINECS: 200-344-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...