DIPPMPO ≥98% (durch TLC, 1H-NMR, 13C-NMR)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-(Diisopropoxyphosphoryl)-5-methyl-1-pyrroline-N-oxide

ALX430119M050
ENZOALX430119M050EA 294 EUR
ENZOALX430119M050
DIPPMPO ≥98% (durch TLC, 1H-NMR, 13C-NMR)
DIPPMPO

Most efficient spin trap for the in vitro and in vivo detection of O-, N-, S-, and C-centered free radicals.


DIPPMPO has a longer life-time than DMPO. More lipophilic (Kp=2.1) than DEPMPO.


Very hygroscopic and soluble in water, 100% ethanol or methylene chloride.

Formel: C₁₁H₂₂NO₄P
Molecular Weight: 263,27 g/mol
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
MDL: MFCD08702711
CAS-Nummer: 527704-58-1

Bestellen

Spezifikation Testergebnisse

Purity (TLC, ¹H- and ¹³C-NMR) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR