NOR-5 ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: (±)-2-((E)-4-Ethyl-3[(Z)-hydroxyimino]6-methyl-5-nitro-heptenyl)-3-pyridinecarboxamide

ALX430125M010
ENZOALX430125M010EA 352 EUR
ENZOALX430125M010
NOR-5 ≥98% (durch HPLC)
NOR-5

Nitric oxide (NO) donor.


The half-life time of 0.5 mM NOR-5 is 20 hours at 37 °C in 0.1 M PBS, pH 7.4.


Soluble in DMSO (10 mg/ml).

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR