IM-54 ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-Methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-1H-pyrrole-2,5-dione

ALX430137M001 ALX430137M005
ENZOALX430137M001EA 53.9 EUR
ENZOALX430137M001 ENZOALX430137M005
IM-54 ≥98% (durch HPLC)
IM-54

Cell permeable inhibitor of necrosis. Selectively blocks oxidative stress-induced necrotic cell death (~3 µm IM-54 prevented ~50% cell death in HL60 cells exposed to 100 µm H2O2). Does not protect against etoposide-induced apoptosis or display antioxidant properties. Does not affect S6K1 and PKC kinase activities at concentrations of 50 µm.

Formel: C₁₉H₂₃N₃O₂
Molecular Weight: 325.40 g/mol
MDL: MFCD18382109
CAS-Nummer: 861891-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR