RITA ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(5,5'-(Furan-2,5-diyl)bis(thiophene-5,2-diyl))dimethanol
ALX430151M005
ENZOALX430151M005EA
638
EUR
ENZOALX430151M005
RITA ≥98%
RITA
Inhibitor of p53/HDM2 interaction
Inhibitor of p53/HDM2 interaction. Activator of p53 function in tumors. Potently inhibits the growth of a variety of cancer cell lines in the submicromolar range. It has been suggested that RITA selectively targets cells expressing p53 by other mechanisms than direct binding to the N-terminal domain of p53.
Formel:
C₁₄H₁₂O₃S₂ Molecular Weight: 292,367 g/mol |
MDL:
MFCD03235294 CAS-Nummer: 213261-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...