4EGI-1 ≥95%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
α-[2-[4-(3,4-Dichlorophenyl)-2-thiazolyl]hydrazinylidene]-2-nitro-benzenepropanoic acid, 2-(2-(4-(3,4-Dichlorophenyl)thiazol-2-yl)hydrazono)-3-(2-nitrophenyl)propanoic acid
ALX430155M002
ENZOALX430155M002EA
332
EUR
ENZOALX430155M002
4EGI-1 ≥95%
4EGI-1
Selective and cell permeable inhibitor of the interaction between the human eukaryotic translation initiation factors 4E (eIF4E) and 4G (eIF4G).
- eIF inhibitor
Brown solid.
Soluble in DMSO.
Formel:
C₁₈H₁₂Cl₂N₄O₄S Molecular Weight: 451,3 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD28142689 CAS-Nummer: 315706-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...