(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%

Lieferant: Enzo Life Sciences

ALX430156M001 ALX430156M005
ENZOALX430156M001EA 99 EUR
ENZOALX430156M001 ENZOALX430156M005
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime

Inibitor of phosphoinositide-dependent kinase 1 and GSK


Inhibitor of phosphoinositide-dependent kinase 1 (PDK1). Potent, reversible and ATP-competitive inhibitor of glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) (IC50=5nM).

Formel: C₁₆H₁₀BrN₃O₂
MDL: MFCD08705318
CAS-Nummer: 667463-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR