Me-7BIO ≥98%

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: (2’Z,3’E)-7-Bromo-1-methylindirubin-3’-oxime

ALX430157M001
ENZOALX430157M001EA 153 EUR
ENZOALX430157M001
Me-7BIO ≥98%
Me-7BIO

Control compound for 6BIO.


Soluble in DMSO.

Bestellen

Spezifikation Testergebnisse

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR