Lamotrigin ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

ALX430160M025 ALX430160M100 ALX430160M500
ENZOALX430160M025EA 245 EUR
ENZOALX430160M025 ENZOALX430160M100 ENZOALX430160M500
Lamotrigin ≥98%
Lamotrigine

Na⁺ channel blocker


Lamotrigine blocks voltage-gated sodium channels. Inhibits the release of glutamate. Neuroprotective. Anticonvulsant.

Formel: C₉H₇Cl₂N₅
Molecular Weight: 256.09 g/mol
MDL: MFCD00865333
CAS-Nummer: 84057-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR