L-Biopterin ≥98%

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2-Amino-6-((1R,2S)-1,2-dihydroxypropyl)pteridin-4(1H)-one, 6-Biopterin

ALX440008M100
ENZOALX440008M100EA 355 EUR
ENZOALX440008M100
L-Biopterin ≥98%
L-Biopterin

Soluble in 1 N sodium hydroxide or 1 N hydrogen chloride; very slightly soluble in water (0.7 mg/ml) (ultrasonication). To obtain concentrated solutions, ammonium hydroxide should be added.

Formel: C₉H₁₁N₅O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00036787
CAS-Nummer: 22150-76-1
Merck Index: 14,01229

Bestellen

Spezifikation Testergebnisse

Purity (assay) ≥98%
Identity Determined by GC-MS
Appearance White to pale yellow powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR