Aminopterin
Lieferant: ENZO LIFE SCIENCES
Synonyme:
4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid, 2-(4-((2,4-Diaminopteridin-6-yl)methylamino)-benzamido)pentanedioic acid, (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, 4-Amino-PGA
ALX440041M050
ENZOALX440041M050EA
103
EUR
ENZOALX440041M050
Aminopterin
Aminopterin
Folic acid antagonist. Potent dihydrofolate reductase inhibitor.
Formel:
C₁₉H₂₀N₈O₅ Molecular Weight: 440,42 g/mol |
MDL:
MFCD00075621 CAS-Nummer: 54-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...