Ionomycin freie Säure
Lieferant: ENZO LIFE SCIENCES
ALX450006M001
ALX450006M005
ENZOALX450006M001EA
149
EUR
ENZOALX450006M001
ENZOALX450006M005
Ionomycin freie Säure
Ionomycin
Formel:
C₄₁H₇₂O₉ Molecular Weight: 709.02 g/mol |
MDL:
MFCD06798385 CAS-Nummer: 56092-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...