D(+)-Biotin
Lieferant: Enzo Life Sciences
ALX460002G001
ALX460002G005
ENZOALX460002G001EA
50.6
EUR
ENZOALX460002G001
ENZOALX460002G005
D(+)-Biotin
D(+)-Biotin
Formel:
C₁₀H₁₆N₂O₃S Schmelzpunkt: 232…233 °C Dichte: 1,41 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005541 CAS-Nummer: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...