Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3) ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione
ALX460007G010
ENZOALX460007G010EA
50.6
EUR
ENZOALX460007G010
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3) ≥98% (durch HPLC)
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Cytotoxic in various cell lines. Induces apoptosis via elevation of peroxide and superoxide radical levels. Affects gap-junctional intercellular communication by mediation of tyrosine phosphorylation.Inhibitor of indoleamine 2,3-dioxygenase (IDO) with an IC50 of ~1µM.
- IDO inhibitor
Off-white to yellow crystalline solid.
Soluble in 96% ethanol (16mg/ml) or DMSO (1mg/ml); almost insoluble in water.
Formel:
C₁₁H₈O₂ Molecular Weight: 172,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001681 CAS-Nummer: 58-27-5 EINECS: 200-372-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...