Alfacalcidol ≥97%

Lieferant: ENZO LIFE SCIENCES

Synonyme: (1R,3S,Z)-5-((E)-2-((1R,3aS,7aR)-7a-Methyl-1-((R)-6-methylheptan-2-yl)hexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha-Hydroxyvitamin D3, 1alpha-Hydroxycholecalciferol

ALX460036M001
ENZOALX460036M001EA 141 EUR
ENZOALX460036M001
Alfacalcidol ≥97%
Alfacalcidol

Vitamin D3 precursor


Converts to active calcitriol (1alpha,25-dihydroxyvitamin D3; Prod. No. BML-DM200) in vivo. Inhibits bone resorption.

Formel: C₂₇H₄₄O₂
Molecular Weight: 400,65 g/mol
MDL: MFCD00868328
CAS-Nummer: 41294-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR