ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)

Lieferant: Enzo Life Sciences

ALX480003G001 ALX480003M050 ALX480003M250
ENZOALX480003G001EA 290 EUR
ENZOALX480003G001 ENZOALX480003M050 ENZOALX480003M250
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Formel: C₂₁H₂₆N₇Na₂O₁₇P₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00065390
CAS-Nummer: 24292-60-2
EINECS: 246-129-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR