6-(Benzyloxy)-7H-purin-2-amin ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
O6-Benzylguanine
ALX480019M010
ENZOALX480019M010EA
83.6
EUR
ENZOALX480019M010
6-(Benzyloxy)-7H-purin-2-amin ≥98% (durch TLC)
6-(Benzyloxy)-7H-purin-2-amine
DNA alkyltransferase substrate and inhibitor
Effective substrate and efficient inactivator of DNA alkyltransferase.
Formel:
C12H11N5O |
MDL:
MFCD00269931 CAS-Nummer: 19916-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...