Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat

Lieferant: ENZO LIFE SCIENCES

Synonyme: (((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate, 5'-ATP·2Na·3H2O, Adenosine-5'-triphosphoric acid disodium salt trihydrate, ATP, 2Na 3H2O, ATP disodium trihydrate

ALX480021G005
ENZOALX480021G005EA 88 EUR
ENZOALX480021G005
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat
Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
Molecular Weight: 605,19 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00150754
CAS-Nummer: 51963-61-2
EINECS: 213-579-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR