8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX480025M001
ENZOALX480025M001EA
371
EUR
ENZOALX480025M001
8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt ≥98% (durch HPLC)
8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt
PKA activator.
Metabolically resistant activator of cAMP-dependent protein kinase (PKA) (cAMP agonist).
No metabolic side effects as observed with dibutyryl-cAMP (N⁶,O²'-dibutyryladenosine-3',5'-cyclic monophosphate, or 8-Br-cAMP (8-bromoadenosine-3',5'-cyclic monophosphate. Significantly more lipophilic and membrane permeant as compared to Sp-cAMPS (adenosine-3',5'-cyclic monophosphothioate, Sp isomer).
Soluble in water or aqueous buffer.
Formel:
C₁₀H₁₀BrN₅O₅PS·Na Molecular Weight: 446,2 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
127634-20-2 |
Spezifikation Testergebnisse
Purity (HPLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...