Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥90% (durch HPLC) K+ channel activator

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: Ado-5′-PPP[S], ATPγ-S . 4Li, Adenosine 5′-[γ-thio]triphosphate tetralithium salt, Adenosine 5′-(3-thiotriphosphate) tetralithium salt

ALX480066M005
ENZOALX480066M005EA 165 EUR
ENZOALX480066M005
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥90% (durch HPLC) K+ channel activator
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt

ATP analog that can substitute ATP in various kinase reactions. Substrate and inhibitor of ATP-dependent enzyme systems. The resulting thiophosphorylated proteins are resistant to dephosphorylation by protein phosphatases. Increases the activity of Ca2 -activated K channels.


Appearance: White to off-white solid
Solubility: Soluble in water (25mg/ml)
Shipping: Ambient
Long Term Storage: -20°C

Formel: C₁₀H₁₂N₅O₁₂PS·4Li
Molecular Weight: 546,98 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077293
CAS-Nummer: 93839-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR