Guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX480072M001
ENZOALX480072M001EA 279 EUR
ENZOALX480072M001
Guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥98% (durch HPLC)
Guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt

Inhibitor of cGMP-dependent protein kinase (PKG) 1α (cGMP antagonist). Resistant against mammalian cyclic nucleotid-dependent phosphodiesterases. No metabolic side effects. Low membrane permeability.


  • PKG inhibitor


White to off-white solid.


Soluble in water (125mM), DMSO (2mM), DMF (1mM), methanol (7mM) or PBS, pH 7.4 (125mM).

Formel: C₁₀H₁₁N₅O₆SP·Na
Molecular Weight: 360,3 · 23,0 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 86562-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR