8-Hydroxyguanosine ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 8-OHG

ALX480075M005
ENZOALX480075M005EA 165 EUR
ENZOALX480075M005
8-Hydroxyguanosine ≥98% (durch HPLC)
8-Hydroxyguanosine

Oxidised form of the nucleoside guanosine.


Formed in vivo by the free radical oxidation of RNA molecules, with subsequent degradation and cleavage of the nucleic acid.


Soluble in concentrated NaOH. Once solubilised it can be further diluted and neutralised with water.

Formel: C₁₀H₁₃N₅O₆
Lagertemperatur: Gefrierschrank
CAS-Nummer: 3868-31-3

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥98%
Identity Determined by UV-VIS and ESI-MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR