5-Aza-2'-deoxycytidine ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

ALX480096M005
ENZOALX480096M005EA 384 EUR
ENZOALX480096M005
5-Aza-2'-deoxycytidine ≥98% (durch HPLC)
5-Aza-2'-deoxycytidine

DNA methyltransferase inhibitor


Inhibitor of DNA methyltransferase. Restores mRNA and protein expression of caspase-8 and TRAIL (tumor necrosis factor-related apoptosis inducing ligand) sensitivity of resistant cell lines. Enhances apoptosis induced by HDAC (histone deacetylase) inhibitors.

Formel: C₈H₁₂N₄O₄
Molecular Weight: 228,21 g/mol
MDL: MFCD00043011
CAS-Nummer: 2353-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR