5-Fluoruracil

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2,4-Dihydroxy-5-fluorpyrimidin, Fluoruracil, 5-FU

ALX480099G005
ENZOALX480099G005EA 120 EUR
ENZOALX480099G005
5-Fluoruracil
5-Fluoruracil

Thymidylate synthetase inhibitor


Potent antitumor agent. Inhibits thymidylate synthetase. Induces p53-dependent apoptosis.

Formel: C₄H₃FN₂O₂
Molecular Weight: 130,08 g/mol
MDL: MFCD00006018
CAS-Nummer: 51-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR