Wasser Endotoxin_free, distilled_double

Lieferant: ENZO LIFE SCIENCES

Synonyme: Wasserstoffoxid, Hydrogenoxid

ALX505008LD15
ENZOALX505008LD15EA 56.1 EUR
ENZOALX505008LD15
Wasser Endotoxin_free, distilled_double
Wasser

Sterile, double distilled endotoxin-free water for use with TLRgrade™ or endotoxin-free grade reagents.


Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR