Human recombinant beta-Nerve Growth Factor (from E. coli)
Lieferant: ENZO LIFE SCIENCES
ALX521005C005
ENZOALX521005C005EA 173 EUR
ENZOALX521005C005
Human recombinant beta-Nerve Growth Factor (from E. coli)
Proteine und Peptide
Produced in E. coli. Dimer of two identical subunits (119 aa) linked together by very strong hydrophobic interactions.
Bestellinformation: Quantity: 5 µg (determined by amino acid analysis).